Learn More
SCL/Tal1 Antibody (1E6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-50285AF350
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Spécification
| SCL/Tal1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
| GST-SCL/Tal1 (N-terminal) fusion protein | |
| 0.1 mL | |
| Primary | |
| Zebrafish | |
| Purified |
| Western Blot, Immunofluorescence | |
| 1E6 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6886 | |
| Store at 4°C in the dark. | |
| IgG1 |