Learn More
SDHD Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26519-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The SDHD antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SDHD. This antibody recognizes human antigen. The SDHD antibody has been validated for the following applications: IF, ELISA analysis.
Spécification
| SDHD | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 6392 | |
| IgG |
| Immunofluorescence, ELISA | |
| 167 μg/mL | |
| Rabbit | |
| Primary | |
| Human |