Learn More
Serum Amyloid A (SAA1) Monoclonal Antibody (OTI3D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marque: Origene Technologies CF814033
Description
Serum Amyloid A (SAA1) Monoclonal antibody specifically detects Serum Amyloid A (SAA1) in Human samples. It is validated for ELISASpécification
| Serum Amyloid A (SAA1) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P0DJI8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6288 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| ELISA | |
| OTI3D6 | |
| Unconjugated | |
| SAA1 | |
| amyloid a; Amyloid fibril protein AA; Amyloid protein A; MGC111216; PIG4; SAA; SAA1; Saa-1; Saa2; serum amyloid A; serum amyloid A 1; serum amyloid A 2; serum amyloid A protein; serum amyloid A1; Serum amyloid A1 protein; Serum amyloid A-1 protein; serum amyloid A-like protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); Serum Amyloid-A; TP53I4; tumor protein p53 inducible protein 4 | |
| Human recombinant protein fragment corresponding to amino acids 19-122 of human SAA (NP_954630) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.