missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA530720
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (90%), Xenopus laevis (94%), Pig (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the epsilon member of the sarcoglycan family. Sarcoglycans are transmembrane proteins that are components of the dystrophin-glycoprotein complex, which link the actin cytoskeleton to the extracellular matrix. Unlike other family members which are predominantly expressed in striated muscle, the epsilon sarcoglycan is more broadly expressed. Mutations in this gene are associated with myoclonus-dystonia syndrome. This gene is imprinted, with preferential expression from the paternal allele. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spécification
| SGCE | |
| Polyclonal | |
| Unconjugated | |
| SGCE | |
| dystonia 11, myoclonic; DYT11; EMBL:AAO37579.1, ECO:0000312; EMBL:AAO37579.1}; Epsilon sarcoglycan; Epsilon SG; Epsilon-sarcoglycan; epsilon-sarcoglycan {ECO:0000312; epsilon-SG; epsilon-SG {ECO:0000250; ESG; e-SG; RGD:1303201}; sarcoglycan; sarcoglycan epsilon; sarcoglycan, epsilon; SGCE; sgce {ECO:0000312; UniProtKB:O43556}; UNQ433/PRO840 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20392, 8910 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O43556, O70258 | |
| SGCE | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human SGCE. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu