Learn More
SGTA Antibody (19E8), Allophycocyanin, R&D Systems™
Code produit 18611928 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18611928 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18611928 Fournisseur Novus Biologicals Code fournisseur NBP259426APC
 Article restreint
Expédition estimée: 07-12-2026
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
SGTA Monoclonal specifically detects SGTA in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Antigène SGTA
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 19E8
Conjugué APC
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formule PBS
Alias de gène alphaSGT, Alpha-SGT, protein containing three tetratricopeptide repeats, SGT1, SGThSGT, small glutamine-rich tetratricopeptide repeat (TPR)-containing, small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha, small glutamine-rich tetratricopeptide repeat-containing protein alpha, UBP, Vpu-binding protein
Espèces hôtes Mouse
Immunogène Recombinant human SGTA (1-313aa) purified from E. coli
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Apoptosis, Cell Biology, Cell Cycle and Replication, Signal Transduction
Primaire ou secondaire Primary
Identification génétique (Entrez) 6449
Espèces cibles Human
Contenu et stockage Store at 4C in the dark.
Forme Purified
Isotype IgG1 κ