missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIP-1 Mouse, PE, Clone: 32/SHIP-1, BD Pharmingen™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566009
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SHIP-1 Monoclonal antibody specifically detects SHIP-1 in Mouse, Rat samples. It is validated for Flow cytometry (Intracellular Staining)) .
Spécification
| SHIP-1 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Cell Biology | |
| Mouse, Rat | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| 32/SHIP-1 | |
| PE | |
| Ship1; Ship; Inpp5d; p150Ship; SIP-145; s-SHIP; 7a33 | |
| Mouse p150 [SHIP] aa. 16-135 | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Reseach Use Only.
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu