Learn More
SHOX2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16366-1-AP-20UL
Code nomenclature Nacres: NA.46
 Afficher plus de versions de ce produit
Code produit. 17284606
  • 145.00€ / 20µL
Description
The SHOX2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SHOX2. This antibody recognizes human, mouse antigen. The SHOX2 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
SHOX2
Polyclonal
Unconjugated
20 μL
6474
Liquid
Western Blot, ELISA
500 μg/mL