missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-7/CD328 Antibody (6-434) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-37732
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Spécification
| Siglec-7/CD328 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1 to 4 μg/mL, CyTOF-ready | |
| Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec-7 | |
| Mouse | |
| 75 kDa | |
| 0.1 mg | |
| Cardiovascular Biology | |
| 27036 | |
| Protein A purified | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 6-434 | |
| Unconjugated | |
| PBS (pH 7.4) with 15mM Sodium Azide | |
| SIGLEC7 | |
| Human dendritic cells. | |
| Protein A purified | |
| RUO | |
| Primary | |
| The mouse monoclonal antibody 6-434 recognizes CD328 (Siglec-7), a 75 kDa transmembrane glycoprotein expressed mainly on NK cells, dendritic cells and monocytes. HLDA 8; WS Code ARN-80652 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu