Learn More
Novus Biologicals™ SIRP gamma/CD172g Antibody (KWAR23) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28671-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SIRP gamma/CD172g Monoclonal antibody specifically detects SIRP gamma/CD172g in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| SIRP gamma/CD172g | |
| Monoclonal | |
| Unconjugated | |
| Q9P1W8 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| KWAR23 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma | |
| SIRPg/ CD172g | |
| 100 μg | |
| Cell Cycle and Replication | |
| 55423 | |
| Human | |
| Purified |