missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIVA Antibody, Biotin, Novus Biologicals™
Chicken Polyclonal Antibody
Marque: Novus Biologicals NBP2-41150B
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SIVA Polyclonal antibody specifically detects SIVA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Spécification
| SIVA | |
| Polyclonal | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Biotin | |
| CD27-binding (Siva) protein, CD27BPCD27-binding protein, Siva-1, SIVA1, apoptosis-inducing factor, Siva-2, SIVAapoptosis regulatory protein Siva | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SIVA. The immunogen is located within the first 50 amino acids of SIVA. Amino Acid Squence: AERYSQEVFEKTKRL | |
| 0.1 mL | |
| Apoptosis | |
| 10572 | |
| Store at 4°C in the dark. | |
| IgY |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu