missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLAIN1 Polyclonal antibody specifically detects SLAIN1 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry
Spécification
Spécification
| Antigène | SLAIN1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugué | DyLight 550 |
| Formule | 50mM Sodium Borate |
| Alias de gène | SLAIN motif family, member 1 |
| Espèces hôtes | Goat |
| Immunogène | E. coli-derived recombinant mouse SLAIN1, Met262-Gln483, Accession # Q68FF7 |
| Méthode de purification | Antigen Affinity-purified |
| Quantité | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?