missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA553966
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: CDGDASFTTP EGPKPRSRCS DWASAVEEDE MRTRVNKEMA RYKRKLLIND FGRERKSSSG SSDSKESMST VPADFETDES VLMRRQKQIN YGKNTI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
This gene encodes a protein that binds to the stem-loop structure in replication-dependent histone mRNAs. Histone mRNAs do not contain introns or polyadenylation signals, and are processed by endonucleolytic cleavage. The stem-loop structure is essential for efficient processing but this structure also controls the transport, translation and stability of histone mRNAs. Expression of the protein is regulated during the cell cycle, increasing more than 10-fold during the latter part of G1.
Spécification
| SLBP | |
| Polyclonal | |
| Unconjugated | |
| SLBP | |
| cb157; ele; hairpin binding protein, histone; HBP; histone binding protein; histone RNA hairpin-binding protein; histone stem loop binding protein; histone stem-loop binding protein; histone stem-loop-binding protein; RNA binding protein; sb:cb157; si:dkey-102m7.2; SLBP; Slbp protein; stem-loop (histone) binding protein; stem-loop binding protein; Unknown (protein for MGC:134385) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7884 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14493 | |
| SLBP | |
| Recombinant protein corresponding to Human SLBP. Recombinant protein control fragment (Product #RP-90817). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu