missing translation for 'onlineSavingsMsg'
Learn More

SLC20A1 Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30497922 Shop All Bio Techne Products
Change view
Click to view available options
Quantité:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité Unit Size
30497922 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30497922 Supplier Novus Biologicals Supplier No. NBP338075AF532
 Article restreint
Expédition estimée: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC20A1 Polyclonal antibody specifically detects SLC20A1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigène SLC20A1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugué Alexa Fluor 532
Formule 50mM Sodium Borate
Alias de gène FLJ41426, Gibbon ape leukemia virus receptor 1, Glvr-1, GLVR1DKFZp686J2397, Leukemia virus receptor 1 homolog, Phosphate transporter 1, PiT-1PIT1, sodium-dependent phosphate transporter 1, solute carrier family 20 (phosphate transporter), member 1, Solute carrier family 20 member 1
Espèces hôtes Rabbit
Immunogène Recombinant fusion protein containing a sequence corresponding to amino acids 257-356 of human SLC20A1 (NP_005406.3).,, Sequence:, KIEREIKCSPSESPLMEKKNSLKEDHEETKLSVGDIENKHPVSEVGPATVPLQAVVEERTVSFKLGDLEEAPERERLPSVDLKEETSIDSTVNGAVQLPN
Méthode de purification Affinity purified
Quantité 0.1 mL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 6574
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C in the dark.
Type de produit Antibody
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.