missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A4/ZIP4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-94856-0.1ml
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SLC39A4/ZIP4 Polyclonal antibody specifically detects SLC39A4/ZIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
| SLC39A4/ZIP4 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| acrodermatitis enteropathica, zinc-deficiency type, FLJ20327, MGC74741, solute carrier family 39 (zinc transporter), member 4, Solute carrier family 39 member 4, zinc transporter ZIP4, ZIP-4, ZIP4AEZ, Zrt- and Irt-like protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-327 of human SLC39A4 (NP_570901.2). SPPAGLLSLLTSGQGALDQEALGGLLNTLADRVHCANGPCGKCLSVEDALGLGEPEGSGLPPGPVLEARYVARLSAAAVLYLSNPEGTCEDARAGLWASHADHLLALLESPKALTPGLSWLLQRMQARAAGQTPKTACVDIPQLLEEAVGAGAPGSAGGVLAALLDHVRSGSCFHALPSPQYFVDFVFQQHSSEVPMTLAELSALMQRLGVGREAHSDHSHRHRGASSRDPVPLISSSNSSSVWDTVCLSARDVMAAYGLSEQAGVTPEAWAQLSPALLQQQLSGACTSQSRPPVQDQLSQSERY | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 55630 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu