missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21649-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
SLIT and NTRK-like family 3 (Slitrk3) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk3 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.Spécification
SLITRK3 | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94933, Q810B9 | |
KIAA0848, SLIT and NTRK like protein 3, SLITRK3 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
22865, 310519, 386750 |
Immunoprecipitation, Western Blot | |
Unconjugated | |
SLITRK3 | |
Liquid | |
Slitrk3 | |
SLITRK3 Fusion Protein Ag16352 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |