missing translation for 'onlineSavingsMsg'
Learn More

Smad2, p Ser465, p Ser467 Antibody, Novus Biologicals™

Code produit 18135198 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mg
Conditionnement:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18135198 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18135198 Fournisseur Novus Biologicals Code fournisseur NBP244217

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Specifically detects Smad2/3 in Human samples, and it is validated for ELISA, Western Blotting

Smad2 Polyclonal specifically detects Smad2 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Spécification

Antigène Smad2 (p Ser465, p Ser467)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugué Unconjugated
Dilution Western Blot 1:1000-1:3000, ELISA 1:10000
Formule 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Numéro d’ordre du gène Q15796
Alias de gène hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2
Symboles de gène(s) SMAD2
Espèces hôtes Rabbit
Immunogène Smad2 [p Ser465, p Ser467] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a C-terminal of human Smad2 protein. (Uniprot: Q15796)
Poids moléculaire de l’antigène 52.31 kDa
Méthode de purification Affinity Purified
Quantité 0.1 mg
État réglementaire RUO
Disciplines de recherche Cancer, Cell Cycle and Replication, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primaire ou secondaire Primary
Identification génétique (Entrez) 4087
Spécificité du test Anti-Smad2 pS465 pS467 antibody is directed against the phosphorylated form of human Smad2 protein at the pS465 and pS467 residues. The resultant affinity purified antibody was then cross adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human Smad2. Reactivity with non-phosphorylated human Smad2 is minimal by ELISA and western blot. A BLAST analysis was used to suggest cross-reactivity with Smad2 protein from human, mouse, rat, orangutan, and bovine based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
Espèces cibles Human, Mouse, Rat, Bovine, Orangutan
Contenu et stockage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Afficher plus Afficher moins

For Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.