Description
SMVT/SLC5A6 Polyclonal antibody specifically detects SMVT/SLC5A6 in Human samples. It is validated for Western Blot
Spécification
Spécification
| Antigène | SMVT/SLC5A6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formule | PBS (pH 7.3), 50% glycerol |
| Alias de gène | Na(+)-dependent multivitamin transporter, SMVTNa+-dependent multivitamin transporter, sodium-dependent multivitamin transporter, solute carrier family 5 (sodium-dependent vitamin transporter), member 6, Solute carrier family 5 member 6 |
| Espèces hôtes | Rabbit |
| Immunogène | A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human SLC5A6 (NP_066918.2). TVATVTTLMPLTTFSKPTGLQRFYSLSYLWYSAHNSTTVIVVGLIVSLLTGRMRGRSLNPATIYPVLPKLLSLLPLSCQKRLHCRSYGQDHLDTGLFPEKPRNGVLGDSRDKEAMALDGTAYQGSSSTCILQETSL |
| Méthode de purification | Affinity purified |
| Afficher plus |