Learn More
EIF1A Antibody, Novus Biologicals™
Product Code. 18606682 Shop All Bio Techne Products
Change view
Click to view available options
Quantité:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité Unit Size
18606682 100 μg 100µL
18691955 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18606682 Supplier Novus Biologicals Supplier No. NBP298066100ul
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Rabbit Polyclonal Antibody
EIF1A Polyclonal antibody specifically detects EIF1A in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Antigène EIF1A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Dilution Immunohistochemistry-Paraffin 1:200-1:1000
Formule PBS (pH 7.0)
Alias de gène eIF-1A, EIF1AP1, eIF-4C, EIF4C, eukaryotic translation initiation factor 1A, X chromosome, eukaryotic translation initiation factor 1A, X-chromosomal, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, Eukaryotic translation initiation factor 4C, FLJ45762, Putative eukaryotic translation initiation factor 1A
Espèces hôtes Rabbit
Immunogène Produced in rabbits immunized with E. coli-derived Human EIF1A fragment
Méthode de purification Antigen and protein A Affinity-purified
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 1964
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Type de produit Antibody
Forme Purified
Isotype IgG
Gamme de produits Novus