missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R2/SSTR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Marque: Novus Biologicals NB300-157SS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Somatostatin R2/SSTR2 Polyclonal specifically detects Somatostatin R2/SSTR2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.
Spécification
| Somatostatin R2/SSTR2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:1000, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Electron Microscopy | |
| P30874, P30875, P30680 | |
| SSTR2 | |
| A synthetic peptide made to the C-terminus of rat Somatostatin Receptor 2 [UniProt# P30680] | |
| 0.025 mL | |
| Alzheimers Research, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6752 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antisera |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu