missing translation for 'onlineSavingsMsg'
Learn More

SP100 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody

Marque:  Novus Biologicals NBP2-97407APC

Code nomenclature Nacres: NA.46

Code produit. 18627172

  • 407.00€ / 0.10mL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

SP100 Polyclonal antibody specifically detects SP100 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification

Spécification

SP100
Polyclonal
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
DKFZp686E07254, FLJ00340, FLJ34579, lysp100b, nuclear antigen Sp100, nuclear autoantigen Sp-100, Nuclear dot-associated Sp100 protein, SP100 nuclear antigen, SP100-HMG nuclear autoantigen, Speckled 100 kDa
Produced in rabbits immunized with E. coli-derived Human SP100 fragment.
0.1 mL
Primary
Human
Purified
Immunocytochemistry, Immunohistochemistry (Paraffin)
APC
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
6672
Store at 4C in the dark.
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.