missing translation for 'onlineSavingsMsg'
Learn More

SRC2 Polyclonal Antibody
GREENER_CHOICE

SRC2 Polyclonal Antibody

Marque:  Invitrogen PA1846

Code nomenclature Nacres: NA.46

Code produit. 11589180

  • 496.00€ / 100µg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

SRC2 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY
Spécification

Spécification

SRC2
Polyclonal
Unconjugated
NCOA2
bHLHe75; c-fgr; Class E basic helix-loop-helix protein 75; c-src2; D1Ertd433e; FLJ43153; Glucocorticoid receptor-interacting protein 1; glucocorticoid receptor-interacting protein-1; Grip1; GRIP-1; hTIF2; I79_019839; KAT13C; MGC75096; NCOA2; NCoA-2; Nuclear receptor coactivator 2; p160 steroid receptor coactivator 2; p55c-fgr; p58c-fgr; SRC2; SRC-2; Steroid receptor coactivator 2; TIF 2; Tif2; transcriptional intermediary factor 2
Rabbit
Antigen affinity chromatography
RUO
100762955, 10499
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q15596
NCOA2
Synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1.
100 μg
Primary
Human, Hamster
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.