Learn More
SRGAP1 Antibody (5D10), Novus Biologicals™
Code produit 18463079 Tous les produits Bio Techne Produits
Change view
Click to view available options
:
0.1 mg; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit
18463079 0.1 mg; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18463079 Fournisseur Novus Biologicals Code fournisseur H00057522M07
Mouse Monoclonal Antibody
SRGAP1 Monoclonal antibody specifically detects SRGAP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Spécification
Antigène SRGAP1
Applications Western Blot, ELISA, Immunofluorescence, Sandwich ELISA
Classification Monoclonal
Clone 5D10
Conjugué Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formule In 1x PBS, pH 7.4
Numéro d’ordre du gène NP_065813
Alias de gène ARHGAP13srGAP1, FLJ22166, KIAA1304SLIT-ROBO Rho GTPase-activating protein 1, Rho GTPase-activating protein 13, SLIT-ROBO Rho GTPase activating protein 1
Espèces hôtes Mouse
Immunogène SRGAP1 (NP_065813, 952 a.a. ∽ 1050 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PLDPETIAQDIEETMNTALNELRELERQSTAKHAPDVVLDTLEQVKNSPTPATSTESLSPLHNVALRSSEPQIRRSTSSSSDTMSTFKPMVAPRMGVQL
Méthode de purification IgG purified
Quantité 0.1 mg
État réglementaire RUO
Disciplines de recherche Growth and Development, Neuronal Cell Markers
Primaire ou secondaire Primary
Identification génétique (Entrez) 57522
Espèces cibles Human, Mouse
Contenu et stockage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG2a κ