Learn More
SRY Mouse anti-Human, Clone: OTI1G6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marque: Origene Technologies CF505268
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This intronless gene encodes a transcription factor that is a member of the high mobility group (HMG)-box family of DNA-binding proteins. This protein is the testis-determining factor (TDF), which initiates male sex determination. Mutations in this gene give rise to XY females with gonadal dysgenesis (Swyer syndrome); translocation of part of the Y chromosome containing this gene to the X chromosome causes XX male syndrome.Spécification
| SRY | |
| Monoclonal | |
| Unconjugated | |
| SRY | |
| SRXX1, SRXY1, TDF, TDY | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6736 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1G6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q05066 | |
| SRY | |
| Full length human recombit protein of human SRY produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.