Learn More
SSR1 Monoclonal Antibody (OTI3E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marque: Origene Technologies TA503263
Description
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.Spécification
| SSR1 | |
| Monoclonal | |
| 0.89 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16967, P43307 | |
| SSR1 | |
| Full length human recombit protein of human SSR1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E5 | |
| Unconjugated | |
| SSR1 | |
| TRAPA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 403951, 6745 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.