Learn More
Gilac™ Stainless Steel Trolley for Large Volume Container
Stainless Steel Trolley for Large Volume Container makes it easier to work at breast height. Equipped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
Marque: Gilac™ G111795
Code nomenclature Nacres: JA.01
Informations supplémentaires : Poids : 8.40000kg
Description
- Trolley equiped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
- Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
- Makes it easier to work at breast height.
Spécification
Trolley | |
Stainless Steel | |
100 L Large Volume Container | |
665 mm |
Large Volume Containers | |
125 mm | |
26.2 in. |