missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Staphylococcus aureus PBP2a (Methicillin-resistant) Monoclonal Antibody (AC10), FITC

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA546570
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Staphylococcus aureus PBP2a (Methicillin-resistant) Monoclonal Antibody for ICC/IF
Methicillin-resistant Staphylococcus aureus (MRSA) is a type of bacteria that is resistant to the antibiotic methicillin. The resistance is due to the presence of a protein called penicillin-binding protein 2a (PBP2a), which is encoded by the mecA gene. PBP2a has a low affinity for methicillin and other beta-lactam antibiotics, which allows the bacteria to survive and multiply in the presence of these drugs. A target description for PBP2a would be a transpeptidase enzyme that is responsible for the synthesis of the bacterial cell wall and is a key factor in the resistance of MRSA to beta-lactam antibiotics.
Spécification
| Staphylococcus aureus PBP2a (Methicillin-resistant) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| AC10 | |
| FITC | |
| MRSA | |
| Native Staphylococcus aureus (methicillin resistant). | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu