Learn More
STING/TMEM173 Antibody (STING1/8187R), Novus Biologicals™
Code produit 18600404 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
20 μg
Conditionnement:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18600404 20 μg 20µg
18656364 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18600404 Fournisseur Novus Biologicals Code fournisseur NBP32061720ug
Rabbit Monoclonal Antibody
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Antigène STING/TMEM173
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone STING1/8187R
Concentration 0.2 mg/ml
Conjugué Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Formule 10mM PBS with 0.05% BSA
Alias de gène endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173
Espèces hôtes Rabbit
Immunogène Recombinant fragment (around aa190-290) of human STING/TMEM173 protein (exact sequence is proprietary)
Méthode de purification Protein A or G purified
Quantité 20 μg
État réglementaire RUO
Disciplines de recherche Cancer, Immunology, Innate Immunity
Primaire ou secondaire Primary
Identification génétique (Entrez) 340061
Espèces cibles Human
Contenu et stockage Store at 4&drg;C. Do not freeze.
Forme Purified
Isotype IgG κ