Learn More
STRBP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-94570-0.02ml
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
STRBP Polyclonal antibody specifically detects STRBP in Human samples. It is validated for Western Blot
Spécification
| STRBP | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp434N214, FLJ11307, FLJ14223, FLJ14984, ILF3L, interleukin enhancer binding factor 3-like, MGC21529, MGC3405, p74, spermatid perinuclear RNA binding protein, spermatid perinuclear RNA-binding protein, SPNRFLJ21960 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-190 of human STRBP (NP_060857.2). CALKHVSDWLDETNKGTKTEGETEVKKDEAGENYSKDQGGRTLCGVMRIGLVAKGLLIKDDMDLELVLMCKDKPTETLLNTVKDNLPIQIQKLTEEKYQVEQCVNEASIIIRNTKEPTLTLKVILTSPLIRDELEKKDGENVSMKDPPDLL | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55342 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |