Learn More
Invitrogen™ Streptococcus Group A Monoclonal Antibody (HSA12-322.2), Invitrogen™
Mouse Monoclonal Antibody
Marque:  Invitrogen™ MA5-46096
Code nomenclature Nacres: NA.46
Code produit. 17890146
  • 424.00€ / 100µg
Description
MA1-10699 detects Streptococci Group A. MA1-10699 has been successfully used in ELISA applications. The MA1-10699 immunogen is UV-inactivated Group A Streptococcus cells.
The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Streptococcus Group A
Monoclonal
0.9 mg/mL
PBS with no preservative; pH 7.4
UV-inactivated Group A Streptococcus cells.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA
HSA12-322.2
Unconjugated
Mouse
Protein G
RUO
Bacteria
Antibody
IgG1