missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1) Antibody (OTI6B11), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-74410V
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SUMO Activating Enzyme E1 (SAE1) Monoclonal specifically detects SUMO Activating Enzyme E1 (SAE1) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| SUMO Activating Enzyme E1 (SAE1) | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SAE1 | |
| Full length recombinant protein of human SUMO Activating Enzyme E1 (SAE1) (NP_005491) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI6B11 | |
| Western Blot, Immunohistochemistry | |
| AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific, Stem Cell Markers | |
| 10055.0 | |
| Store at 4°C in the dark. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu