Learn More
Superoxide Dismutase 1 (SOD1) (Antioxidant Enzyme) Mouse, Clone: SOD1/2089, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio 6647-MSM1-P1
Code nomenclature Nacres: NA.46
Code produit. 15980878
  • ONLINE EXCLUSIVE PRICE
    554.40€
    478.00€ / 100µg
    Économisez 76.40€
    13% de réduction
Spécification
Superoxide Dismutase 1 (SOD1) (Antioxidant Enzyme)
Cytoplasmic
SOD1/2089
Unconjugated
Mouse
23kDa
Protein A/G purified from Bioreactor concentrate.
RUO
6647
IgG
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant full-length human SOD1 protein
HeLa or Jurkat Cells. Breast or Ovarian carcinoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.