missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SynGAP Recombinant Rabbit Monoclonal Antibody (HL2292)
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA550478
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SynGAP Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Synaptic Ras-GTPase activating protein (SynGAP) is a PDZ-domain interacting protein expressed almost exclusively in the brain. SynGAP has been shown to be a major component of postsynaptic density and associates with post synaptic density 95 kDa (PSD-95), a synaptic scaffold protein, and synaptic N-methyl-D-aspartate (NMDA) receptors. SynGAP Ras-GTPase activity has also been shown to be inhibited by phosphorylation by CaM kinase II, another abundant protein located in the postsynaptic density. It has been proposed that SynGAP may function as a negative regulator of MAP Kinase activation in the absence of active NMDA receptors.
Spécification
| SynGAP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| F6SEU4, Q9QUH6 | |
| SYNGAP1 | |
| Recombinant fragmemt of human SYNGAP1. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2292 | |
| Unconjugated | |
| SYNGAP1 | |
| DKFZp761G1421; Gm1963; KIAA1938; MRD5; Neuronal R; Neuronal RasGAP; OTTHUMP00000209046; p135 SynGAP; Ras GTPase-activating protein SynGAP; ras/Rap GTPase-activating protein SynGAP; RASA1; RASA5; synaptic Ras GTPase activating protein 1; synaptic Ras GTPase activating protein 1 homolog; synaptic Ras GTPase activating protein 1 homolog (rat); synaptic Ras GTPase activating protein, 135kDa; synaptic Ras GTPase-activating protein 1; Synaptic Ras-GAP 1; SYNGAP; Syngap1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 192117, 240057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu