Learn More
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, PE, Clone: GL3, STEMCELL Technologies
Hamster Monoclonal Antibody
Marque:  Stemcell Technologies 60104PE
Encompass_Preferred
Code nomenclature Nacres: NA.46
Code produit. 17149031
  • 303.75€ / 100µg
Description
  • The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens
  • TCRγ/δ comprises a heterodimer (∼ 80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface
  • It is a member of the immunoglobulin superfamily
  • TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues
  • These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses
  • Once activated, they secrete effector cytokines in a subtype- and context-specific manner
  • Most γ/δ T cells are CD4-/CD8-, though some express CD8
  • The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells
Spécification
Spécification
T Cell Receptor Gamma/Delta
Monoclonal
PE
100 μg
Mouse
T Cells, T Cells, Other Subsets
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay
GL3
Hamster
Primary
IgG2 κ
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.