missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Talin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA584087
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: AADLNQSAGE VVHATRGQSG ELAAASGKFS DDFDEFLDAG IEMAGQAQTK EDQIQVIGNL KNISMA Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene encodes a protein related to talin 1, a cytoskeletal protein that plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. This protein has a different pattern of expression compared to talin 1 but, like talin 1, is thought to associate with unique transmembrane receptors to form novel linkages between extracellular matrices and the actin cytoskeleton.
Spécification
| Talin 2 | |
| Polyclonal | |
| Unconjugated | |
| Tln2 | |
| 5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; mKIAA0320; RGD1565416; talin 2; talin-2; TLN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y4G6 | |
| Tln2 | |
| Recombinant protein corresponding to Human TLN2. Recombinant protein control fragment (Product #RP-108205). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu