missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-55934
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TAP1 Polyclonal specifically detects TAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spécification
| TAP1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| ABC transporter, MHC 1, ABCB2FLJ41500, antigen peptide transporter 1, APT1, ATP-binding cassette sub-family B member 2, ATP-binding cassette, sub-family B (MDR/TAP), member 2, D6S114E, Peptide supply factor 1, Peptide transporter involved in antigen processing 1, Peptide transporter PSF1, Peptide transporter TAP1, PSF-1, PSF1ABC17, Really interesting new gene 4 protein, RING4FLJ26666, TAP1*0102N, TAP1N, transporter 1, ATP-binding cassette, sub-family B (MDR/TAP), transporter associated with antigen processing, transporter, ATP-binding cassette, major histocompatibility complex, 1, Y3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| TAP1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DDATSALDANSQLQVEQLLYESPERYSRSVLLITQHLSLVEQADHILFLEGGAIREGGTHQQLMEK | |
| 100 μL | |
| Cell Biology, Diabetes Research, Membrane Trafficking and Chaperones | |
| 6890 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu