Learn More
PeproTech TARC Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Marque: PeproTech 500-P97-1MG
Description
AA Sequence of recombinant protein: ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TARC (CCL17). Anti-Human TARC (CCL17)-specific antibody was purified by affinity chromatography employing an immobilized Human TARC (CCL17) matrix. Sandwich ELISA: To detect Human TARC (CCL17) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TARC (CCL17) (500-P97Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TARC (CCL17). Western Blot: To detect hTARC by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hTARC is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
Spécification
| TARC | |
| Polyclonal | |
| Unconjugated | |
| CCL17 | |
| A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6361 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q92583 | |
| CCL17 | |
| E.coli-derived Recombinant Human TARC (CCL17). | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.