missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBCA Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA557683
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: MRAEDGENYD IKKQAEILQE SRMMIPDCQR RLEAAYLDLQ RILENEKDLE EAEEY Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
The product of this gene is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Spécification
| TBCA | |
| Polyclonal | |
| Unconjugated | |
| TBCA | |
| CFA; chaperonin cofactor A; co-chaperonin associated with a- & b-tubulin; cofactor A; Tbca; Tbca13; TCP1-chaperonin cofactor A; tubulin cofactor a; tubulin folding cofactor A; tubulin-folding cofactor A; Tubulin-specific chaperone A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6902 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75347 | |
| TBCA | |
| Recombinant protein corresponding to Human TBCA. Recombinant protein control fragment (Product #RP-96937). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu