missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12157-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.Spécification
| TCN2 | |
| Polyclonal | |
| Unconjugated | |
| TCN2 | |
| D22S676, D22S750, TC 2, TC II, TC2, TCII, TCN2, Transcobalamin 2, Transcobalamin II | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21452, 64365, 6948 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88968, P20062, Q9R0D6 | |
| Tcn2 | |
| TCN2 Fusion Protein Ag2825 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu