Learn More
TCR V delta 1 Antibody (TS8.2), Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-22506
Code nomenclature Nacres: NA.46
Code produit. 18110012
  • 482.00€ / 200µg
Description
TCR V delta 1 Monoclonal specifically detects TCR V delta 1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
TCR V delta 1
Monoclonal
0.2 mg/mL
Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
TRD
Human TCR V delta 1
200 μg
Primary
This antibody reacts with Human.
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
TS8.2
Unconjugated
T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@
Mouse
Protein G purified
RUO
6964
Human
Ascites
For Research Use Only