missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDO2 Antibody (OTI2A4), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-45995
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
TDO2 Monoclonal specifically detects TDO2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| TDO2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| TDOTO, TPH2EC 1.13.11.11, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase | |
| Mouse | |
| 47.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2A4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| TDO2 | |
| Full length human recombinant protein of human TDO2(NP_005642) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| 6999 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur