missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tampon d’étalonnage technique pH 10, Hanna Instruments™
Click to view available options
Quantité:
20 mL
Spécification
Couleur | Incolore |
pH | 10 |
Forme physique | Liquide |
Quantité | 20 mL |
Type de solution | Buffer |
Formule | Aqueous Buffer Solution |
Odeur | Odorless |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'