Learn More
Invitrogen™ Teplizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA559030
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
This is anti-CD3 monoclonal antibody that is used in high-risk groups to prevent or delay the development of clinical type 1 diabetes (T1D). High-risk groups here refer to the presence of two or more T1D-related autoantibodies in the body.
Spécification
| Teplizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MGA031; MGA-031; PRV-031 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 0.01% Tween 80, 8% sucrose and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |