missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23996-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TMEM192 is a widely expressed, four transmembrane protein that localizes to late endosomal/lysosomal membranes. This localization is mediated by two dileucine motifs present at the amino-terminus of TMEM192. A cysteine residue near the carboxy residue is responsible for disulfide bond formation in TMEM192 homodimers. Little is known about the function of this protein.Spécification
| TFAM | |
| Polyclonal | |
| Unconjugated | |
| TFAM | |
| MtTF1, mtTFA, TCF 6, TCF6, TCF6L1, TCF6L2, TCF6L3, TFAM, Transcription factor 6, Transcription factor 6 like 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7019 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q00059 | |
| TFAM | |
| TFAM Fusion Protein Ag21196 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu