Learn More
TGF-beta 1 Antibody (fresolimumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals NBP3-28822CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| TGF-beta 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| TGFb1 | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 7040 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| fresolimumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |