missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin-1 Antibody (A6.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Marque: Novus Biologicals NB100-2059
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Thrombospondin-1 Monoclonal antibody specifically detects Thrombospondin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Block/Neutralize.
Spécification
| Thrombospondin-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50, Immunohistochemistry-Frozen, Blocking Assay | |
| P07996 | |
| THBS1 | |
| BALB/C mice were injected with reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets. | |
| 0.25 mL | |
| Cancer | |
| 7057 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Sheep | |
| Purified |
| Blocking Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| A6.1 | |
| Unconjugated | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| THBS, THBS-1, thrombospondin 1, thrombospondin-1p180, TSP1thrombospondin-1, TSPTSP-1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This is specific to 450 kD protein, a non-reduced form, and 170 to 180 kD protein, a reduced form, of Thrombospondin. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu