Learn More
Thymine Dimer Antibody (H3) - BSA Free, Novus Biologicals™
Code produit 18736913 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.025 mL
Conditionnement:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18736913 0.025 mL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18736913 Fournisseur Novus Biologicals Code fournisseur NB6001141
Mouse Monoclonal Antibody
Thymine Dimer Monoclonal antibody specifically detects Thymine Dimer in Human, Nonspecies specific, Chicken samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot, In Situ Hybridization (ISH).
TRUSTED_SUSTAINABILITY
Spécification
Antigène Thymine Dimer
Applications ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot, Southern Blot
Classification Monoclonal
Clone H3
Concentration 2.0 mg/mL
Conjugué Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Dot Blot 0.5-1 ug/ml, Southern Blot 0.5-1 ug/ml
Espèces hôtes Mouse
Immunogène tetra nucleotide containing cyclobutane thymine dimer (GTTG) conjugated to chicken gamma globulin.
Méthode de purification Protein A or G purified
Quantité 0.025 mL
État réglementaire RUO
Disciplines de recherche DNA Repair
Primaire ou secondaire Primary
Spécificité du test Monoclonal Anti-Thymine Dimer reacts specifically with the (5'-6') cyclobutane type of homothymine or thyminecytosine heterodimers. The antibody reacts with thymine dimers in single-stranded DNA, and has a lower affinity for the dimer in short oligonucleotides (a tail of minimum 10-20 thymine residues is required for efficient labeling of oligonucleotide probes). The product enables a sensitive and non-radioactive method for labeling, detection, and quantification of DNA fragments using ELISA, competitive ELISA, immunocytochemistry (laser-scan microscopy) and Southern immunoblotting.
Espèces cibles Human, Non-species specific, Chicken
Contenu et stockage Store at -20C. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG1
Afficher plus Afficher moins
For Research Use Only