Learn More
TIMP1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen 500-P280BT-1MG
Description
AA Sequence of recombinant protein: CTCVPPHPQT AFCNSDLVIR AKFVGTPEVN QTTLYQRYEI KMTKMYKGFQ ALGDAADIRF VYTPAMESVC GYFHRSHNRS EEFLIAGKLQ DGLLHITTCS FVAPWNSLSL AQRRGFTKTY TVGCEECTVF PCLSIPCKLQ SGTHCLWTDQ LLQGSEKGFQ SRHLACLPRE PGLCTWQSLR SQIA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TIMP-1. Anti-Human TIMP-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hTIMP-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human TIMP-1 (500-P280) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hTIMP-1. Western Blot: To detect hTIMP-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hTIMP-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P280BT-1MG will be provided as 2 x 500 μg
TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.
Spécification
| TIMP1 | |
| Polyclonal | |
| Biotin | |
| TIMP1 | |
| CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7076 | |
| -20°C |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P01033 | |
| TIMP1 | |
| E.coli-derived, 20.6 kDa Recombinant Human TIMP-1 | |
| 2 x 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.