Learn More
TIMP2 (Tissue Inhibitor of Metalloproteinase 2) Mouse, Clone: TIMP2/2044, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 7077-MSM4-P0
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| TIMP2 (Tissue Inhibitor of Metalloproteinase 2) | |
| Cytoplasm and Cell Surface | |
| TIMP2/2044 | |
| Unconjugated | |
| Mouse | |
| 21kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 7077 | |
| IgG1 κ |
| Immunohistochemistry, ELISA | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human TIMP2 protein fragment (aa 48-220) | |
| HeLa cells. Normal Placenta. Breast, Colon, Endometrial, Prostate or Ovarian Carcinoma. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |