Learn More
TIMP2 (Tissue Inhibitor of Metalloproteinase 2) Mouse, Clone: TIMP2/2044, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio 7077-MSM4-P1
Code nomenclature Nacres: NA.46
Code produit. 15980978
  • ONLINE EXCLUSIVE PRICE
    554.40€
    478.00€ / 100µg
    Économisez 76.40€
    13% de réduction
Spécification
TIMP2 (Tissue Inhibitor of Metalloproteinase 2)
Cytoplasm and Cell Surface
TIMP2/2044
Unconjugated
Mouse
21kDa
Protein A/G purified from Bioreactor concentrate.
RUO
7077
IgG1 κ
Immunohistochemistry, ELISA
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human TIMP2 protein fragment (aa 48-220)
HeLa cells. Normal Placenta. Breast, Colon, Endometrial, Prostate or Ovarian Carcinoma.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.