missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1 Antibody (TLE1/2062), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-79914APC
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TLE1 Monoclonal antibody specifically detects TLE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Spécification
| TLE1 | |
| Monoclonal | |
| APC | |
| E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1 | |
| Recombinant human TLE1 fragment (aa 175-338) (exact sequence is proprietary) (Uniprot: Q04724) | |
| 0.1 mL | |
| Signal Transduction | |
| 7088 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| TLE1/2062 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction